Please login to your online account to display your discounted pricing

AM 404, Tocris

Anandamide transport inhibitor

$116.01 - $486.49

Chemical Identifiers

Chemical Name or Material AM 404
Assay Percent Range >98%
Molecular Formula C26H37NO2
Formula Weight 395.58
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

111610

 
r & d systems
RDS111610
10mg Each for $116.01

111650

 
r & d systems
RDS111650
50mg Each for $486.49
Description & Specifications

Specifications

Formula Weight 395.58
Quantity 10mg
Chemical Name or Material AM 404
Molecular Formula C26H37NO2
Assay Percent Range >98%
CAS 198022-70-7
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C