Please login to your online account to display your discounted pricing

AM 404 (in Tocrisolve 100), Tocris

Anandamide transport inhibitor (in water-soluble emulsion)

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material AM 404 (in Tocrisolve™ 100)
Assay Percent Range >98%
Molecular Formula C26H37NO2
Formula Weight 395.58
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

16855

  SDS
r & d systems
RDS16855
5mg Each for $139.08

168525

 
r & d systems
RDS168525
25mg Each for $585.69
Description & Specifications

Specifications

Formula Weight 395.58
Quantity 5mg
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
Recommended Storage Store at +4°C
Molecular Formula C26H37NO2
CAS 198022-70-7
Chemical Name or Material AM 404 (in Tocrisolve™ 100)
Assay Percent Range >98%
Safety and Handling

Store at +4°C