Please login to your online account to display your discounted pricing

Alfa Aesar™ AM-404, 98%


Quantity 10mg

Manufacturer: alfa aesar™  J67201MA

 View more versions of this product

Catalog No. AAJ67201MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 395.59
Chemical Name or Material AM-404
Physical Form Solid
Assay 98%
MDL Number MFCD03095721
CAS 198022-70-7
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z-eicosatetraenamide; N-(4-Hydroxyphenyl)arachidonoylamide
Sensitivity Air Sensitive
Color Waxy
Quantity 10mg

Specifications

Chemical Name or Material AM-404
CAS 198022-70-7
Formula Weight 395.59
MDL Number MFCD03095721
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z-eicosatetraenamide; N-(4-Hydroxyphenyl)arachidonoylamide