Please login to your online account to display your discounted pricing

Alfa Aesar™ AM-404, 98%

Chemical Identifiers

Chemical Name or Material AM-404
CAS 198022-70-7
Formula Weight 395.59
MDL Number MFCD03095721
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z-eicosatetraenamide; N-(4-Hydroxyphenyl)arachidonoylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67201MA

 
alfa aesar™
J67201MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 395.59
Chemical Name or Material AM-404
Physical Form Solid
Assay 98%
MDL Number MFCD03095721
CAS 198022-70-7
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z-eicosatetraenamide; N-(4-Hydroxyphenyl)arachidonoylamide
Sensitivity Air Sensitive
Color Waxy
Quantity 10mg