Please login to your online account to display your discounted pricing

AM 251, Tocris


Potent CB1 antagonist; Also GPR55 agonist

Quantity 10mg

Manufacturer: r & d systems  111710

 View more versions of this product

Catalog No. 111710

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 555.24
Quantity 10mg
CAS 183232-66-8
Recommended Storage Store at Room Temperature
Chemical Name or Material AM 251
Assay Percent Range >99%
Synonym N-(Piperidin-1-yl)-5-(4-iodophenyl)-1-(2, 4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
Molecular Formula C22H21Cl2IN4O

Specifications

Chemical Name or Material AM 251
Assay Percent Range >99%
Molecular Formula C22H21Cl2IN4O
Formula Weight 555.24
Synonym N-(Piperidin-1-yl)-5-(4-iodophenyl)-1-(2, 4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
Accessories

Store at Room Temperature