Please login to your online account to display your discounted pricing

AM 114, Tocris

20S proteasome inhibitor

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material AM 114
Assay Percent Range >97%
Molecular Formula C20H21B2NO5
Formula Weight 377.01
Synonym 3, 5-Bis-[benzylidene-4-boronic acid]-1-methylpiperidin-4-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

256450

 
r & d systems
RDS256450
50mg Each for $762.58

256410

 
r & d systems
RDS256410
10mg Each for $181.19
Description & Specifications

Specifications

Formula Weight 377.01
Quantity 50mg
Chemical Name or Material AM 114
Assay Percent Range >97%
Molecular Formula C20H21B2NO5
Synonym 3, 5-Bis-[benzylidene-4-boronic acid]-1-methylpiperidin-4-one
Recommended Storage Store at +4°C
CAS 856849-35-9
Safety and Handling

Store at +4°C