missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Alytesin

For cellular and molecular biology applications

$65.18 - $236.39

Specifications

Product Type Alytesin
Molecular Weight 1535.79
Storage Requirements -30° to -10°C
Sequence Glp-Gly-Arg-Leu-Gly-Thr-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66018MCR
View Documents
Alfa Aesar™
J66018MCR
1mg Each for $65.18
 
AAJ66018LB0
View Documents
Alfa Aesar™
J66018LB0
5mg Each for $236.39
 
Specifications

Specifications

Alytesin
-30° to -10°C
Solid
HPLC
1535.79
Glp-Gly-Arg-Leu-Gly-Thr-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2
RUO

For Research Use Only