missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum wire, 99.999% (metals basis), Puratronic™, Alfa Aesar™

Form: approx. 2.1g/m; Temper: soft

Manufacturer:  Thermo Fisher Scientific 10747H8

 View more versions of this product

Catalog No. AA10747H8



Specifications

Specifications

Aluminum
Annealed
Silver-colored wire
Al
adom
[Al]
Odorless
5359268
Wire, 1.0mm (0.04 in.) dia.
300m
7429-90-5
99.999% (Metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
Silver

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

EINECSNumber : 231-072-3

Documents