Please login to your online account to display your discounted pricing

Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.999% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Aluminum wire
CAS 7429-90-5
Name Note 1.0mm (0.04 in.) dia, annealed, Puratronic™
MDL Number MFCD00134029
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA10747G5

  SDS
alfa aesar
10747G5
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Temper: Soft
Quantity 5m
MDL Number MFCD00134029
TSCA TSCA
CAS 7429-90-5
Odor Odorless
Chemical Name or Material Aluminum wire
Density ≈2.1g/m
Assay 99.999%
Assay Percent Notes (metals basis)
Name Note 1.0mm (0.04 in.) dia, annealed, Puratronic™