missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum wire, 0.81mm (0.032 in.) dia., annealed, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 43336H2

 View more versions of this product

Catalog No. AA43336H2



Specifications

Specifications

Aluminum wire
50m
7429-90-5
Al
adom
[Al]
Odorless
5359268
Wire
Puratronic™
≈1.4g/m
0.81mm (0.032 in.) dia., Annealed, Temper: soft
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99.999%

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents