missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Wire, 0.5mm (0.02 in.) dia, annealed, Puratronic™, 99.999% (Metals basis), Alfa Aesar™

Form: Approx. 0.53g/m; Temper: soft

Manufacturer:  Thermo Fisher Scientific 10748H7

 View more versions of this product

Catalog No. AA10748H7



Specifications

Specifications

Aluminum
Annealed
Silver-colored wire
Al
adom, alumina fibre, aluminio, aluminium, aluminium flake, dust, metal, metana, noral aluminium, powder
[Al]
Odorless
5359268
Wire, ≈0.53g/m,Temper: soft
250m
7429-90-5
99.999% (Metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
Silver

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom, alumina fibre, aluminio, aluminium, aluminium flake, dust, metal, metana, noral aluminium, powder
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents