missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum wire, 0.38mm (0.015 in.) dia., 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 45100H2

 View more versions of this product

Catalog No. AA45100H2



Specifications

Specifications

Aluminum wire
50m
7429-90-5
Al
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
Odorless
aluminum
5359268
Wire
≈0.31g/m
0.38mm (0.015 in.) dia.
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
[Al]
26.982
CHEBI:33629
99.99%

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents