missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum wire, 0.152mm (0.006 in.) dia., hard, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 44579G5

 View more versions of this product

Catalog No. AA44579G5



Specifications

Specifications

Aluminum wire
5m
7429-90-5
Al
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
Odorless
aluminum
5359268
Wire
Puratronic™
≈0.049g/m
0.152mm (0.006 in.) dia., Hard, Temper: as drawn
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
[Al]
26.982
CHEBI:33629
99.999%

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents