missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum wire, 0.152mm (0.006in) dia, hard, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
$149.48 - $368.07
Chemical Identifiers
CAS | 7429-90-5 |
---|---|
Molecular Formula | Al |
Molecular Weight (g/mol) | 26.98 |
MDL Number | MFCD00134029 |
InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
PubChem CID | 5359268 |
ChEBI | CHEBI:33629 |
IUPAC Name | aluminum |
SMILES | [Al] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7429-90-5 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
aluminum |
Al | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
CHEBI:33629 | |
[Al] |
Specifications
7429-90-5 | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
[Al] | |
26.98 | |
CHEBI:33629 | |
Puratronic™ | |
≈0.049g/m | |
Aluminum wire, 0.152mm (0.006 in.) dia., Hard, Temper: as drawn |
Al | |
5 m | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminum | |
5359268 | |
99.999% | |
Odorless | |
Wire | |
(metals basis) |
Safety and Handling
EINECSNumber : 231-072-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only