missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum wire, 0.076mm (0.003 in.) dia., hard, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 44577G9

 View more versions of this product

Catalog No. AA44577G9



Specifications

Specifications

Aluminum wire
25m
7429-90-5
Al
adom
[Al]
Odorless
5359268
Wire, ≈0.012g/m,Temper: as drawn
Puratronic™
≈0.012g/m
0.076mm (0.003 in.) dia., Hard, Temper: as drawn
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99.999%

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents