missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Wire, 0.05mm (0.002 in.) dia, hard, 99.99% (Metals basis), Alfa Aesar™

Form: Approx. 0.0053g/m; Temper: as drawn

Chemical Identifiers

CAS 7429-90-5
Physical Form Wire, 0.05mm (0.002 in.) dia.
Assay Percent Range 99.99% (Metals basis)
MDL Number MFCD00134029
Chemical Name or Material Aluminum, Hard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40676G1 SDS Alfa Aesar
40676G1
1m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA40676G5 SDS Alfa Aesar
40676G5
5m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA40676G6 SDS Alfa Aesar
40676G6
10m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7429-90-5
99.99% (Metals basis)
Aluminum, Hard
Wire, 0.05mm (0.002 in.) dia.
MFCD00134029

Specifications

1m
Silver-colored wire
Wire, 0.05mm (0.002 in.) dia.
MFCD00134029
Temper: as drawn
Aluminum, Hard
7429-90-5
2.7g/cm3
99.99% (Metals basis)
Odorless
Silver