Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum titanium oxide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Aluminum titanium oxide
CAS 12004-39-6
Formula Weight 181.86
MDL Number MFCD00064749
Synonym Aluminum titanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1448414

  SDS
alfa aesar™
1448414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1448422

  SDS
alfa aesar™
1448422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.86
Odor Odorless
Health Hazard 2 GHS H Statement
H335
May cause respiratory irritation.
CAS 12004-39-6
Synonym Aluminum titanate
Health Hazard 3 GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay Percent Notes (metals basis)
MDL Number MFCD00064749
Physical Form Powder
Health Hazard 1 Warning
Quantity 25g
Chemical Name or Material Aluminum titanium oxide
Mesh Size −100 Mesh
Assay 99.5%
Safety and Handling

Warning

GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H335
May cause respiratory irritation.