missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum titanium oxide, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 1448422

 View more versions of this product

Catalog No. AA1448422



Specifications

Specifications

Aluminum titanium oxide
(metals basis)
MFCD00064749
CNRZQDQNVUKEJG-UHFFFAOYSA-N
oxo-bis(oxoalumanyloxy)titanium
16213786
Powder
−100 Mesh
100g
12004-39-6
Al2O5Ti
aluminum titanate, powder, oxo-bis oxoalumanyloxy titanium, trioxo-2,4-dioxa-1,5-dialumina-3-titanapentane, aluminum titanate, nanopowder, <25 nm particle size bet trace metals basis
O=[Al]O[Ti](=O)O[Al]=O
181.825
181.86
99.5%
Odorless

Chemical Identifiers

12004-39-6
181.825
CNRZQDQNVUKEJG-UHFFFAOYSA-N
16213786
O=[Al]O[Ti](=O)O[Al]=O
Al2O5Ti
MFCD00064749
aluminum titanate, powder, oxo-bis oxoalumanyloxy titanium, trioxo-2,4-dioxa-1,5-dialumina-3-titanapentane, aluminum titanate, nanopowder, <25 nm particle size bet trace metals basis
oxo-bis(oxoalumanyloxy)titanium
Safety and Handling

Safety and Handling

GHS H Statement
H335
May cause respiratory irritation.

GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents