missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum sulfide, 99+%, (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01285114

 View more versions of this product

Catalog No. AA1285114



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aluminum sulfide
Yellow
Pieces
>99% (Metal basis)
MFCD00014162
[Al+3].[Al+3].[S--].[S--].[S--]
150.14
Like rotten eggs
Yellowish granular
1100°C
1302-81-4
25 g
Al2S3
COOGPNLGKIHLSK-UHFFFAOYSA-N
dialuminium(3+) trisulfanediide
150.16
Moisture sensitive
2.02

Chemical Identifiers

1302-81-4
150.14
COOGPNLGKIHLSK-UHFFFAOYSA-N
[Al+3].[Al+3].[S--].[S--].[S--]
Al2S3
MFCD00014162
dialuminium(3+) trisulfanediide
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : DOT Class: 4.3, Packaging Group: III

EINECSNumber : 215-109-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only