missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum sulfate hydrate, Puratronic™, 99.999% (metals basis)

Chemical Identifiers

CAS 17927-65-0
Formula Weight 342.15 (Anhydrous)
MDL Number MFCD00149136
Merck Index 14,366
Chemical Name or Material Aluminum sulfate hydrate
Grade Puratronic™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1062906 SDS Alfa Aesar™
1062906
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1062914 SDS Alfa Aesar™
1062914
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

17927-65-0
MFCD00149136
Aluminum sulfate hydrate
342.15 (Anhydrous)
14,366
Puratronic™

Specifications

17927-65-0
342.15 (Anhydrous)
14,366
White
5g
99.999%, (metals basis)
Crystalline
MFCD00149136
1.61g/mL
3.5
Aluminum sulfate hydrate
Puratronic™

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BD1700000

TSCA : TSCA