missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum sulfate hydrate, 97+%, Alfa Aesar

Manufacturer:  Alfa Aesar 1236236

 View more versions of this product

Catalog No. AA1236236



Specifications

Specifications

Aluminum sulfate hydrate
17927-65-0
Al2H2O13S3
White crystalline
BUACSMWVFUNQET-UHFFFAOYSA-H
dialuminum;trisulfate;hydrate
3080712
Crystalline
Odorless
500g
hydrate
>97%
MFCD00149136
aluminium sulfate hydrate, aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o, aluminum sulfate hydrate
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
360.146
342.15
1.61g/cm3
White

Chemical Identifiers

17927-65-0
360.146
BUACSMWVFUNQET-UHFFFAOYSA-H
3080712
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Al2H2O13S3
MFCD00149136
aluminium sulfate hydrate, aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o, aluminum sulfate hydrate
dialuminum;trisulfate;hydrate
Safety and Handling

Safety and Handling

H318-H315-H335

EINECSNumber : 233-135-0

Documents