Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum spheres, 7.94mm (0.31 in.) dia., 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Aluminum
CAS 7429-90-5
Name Note 7.94mm (0.31 in.) dia.
MDL Number MFCD00213972
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42031KP

  SDS
alfa aesar™
42031KP
500 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500 pieces
Avg. Mol. Wt. or Mol. Wt. Range Alloy 1100, Al:Cu; ≥99:0.12 wt%
Assay 99%
Chemical Name or Material Aluminum
Physical Form Spheres
MDL Number MFCD00213972
TSCA TSCA
CAS 7429-90-5
Name Note 7.94mm (0.31 in.) dia.
Assay Percent Notes (metals basis)