missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 4432822

 View more versions of this product

Catalog No. AA4432822



Specifications

Specifications

Aluminum slug
100g
7429-90-5
Al
adom
[Al]
Odorless
5359268
≈0.54g/slug
Puratronic™
≈0.54g/slug
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99.9999%

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents