missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum shot, approx. 4-8mm (0.2-0.3in), Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
approx. 4-8mm (0.2-0.3 in.)
Supplier: Thermo Scientific Chemicals 01057318
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum shot | |
7429-90-5 | |
Al | |
50 g | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminum | |
5359268 | |
26.98 |
shot | |
99.99% (metals basis) | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
[Al] | |
26.98 | |
CHEBI:33629 | |
Sheet |
Chemical Identifiers
7429-90-5 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
[Al] |
Al | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
CHEBI:33629 |
Safety and Handling
EINECSNumber : 231-072-3
RTECSNumber : BD0330000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only