Learn More
Alfa Aesar™ Aluminum selenide, 99% (metals basis)
Manufacturer: Alfa Aesar™ 1313203
Specifications
Aluminum selenide | |
(metals basis) | |
UN3283 | |
unii-4r7pmp996q, aluminum selenide, aluminium selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide | |
290.84 | |
99% | |
Odorless | |
Moisture Sensitive | |
1g |
1302-82-5 | |
MFCD00014158 | |
14,361 | |
Decomposes to Al203 and SeO2 in water | |
Pieces | |
3.437g/mL | |
6.5mm & down | |
Brown |
Chemical Identifiers
1302-82-5 | |
MFCD00014158 | |
unii-4r7pmp996q, aluminum selenide, aluminium selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide |
Safety and Handling
GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
TSCA : TSCA