Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum selenide, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Aluminum selenide
CAS 1302-82-5
Formula Weight 290.84
MDL Number MFCD00014158
UN Number UN3283
Merck Index 14,361
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1313203

  SDS
alfa aesar™
1313203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1313214

  SDS
alfa aesar™
1313214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 290.84
Color Pale brown
Odor Odorless
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
Assay Percent Notes (metals basis)
MDL Number MFCD00014158
Solubility Information Decomposes to Al203 and SeO2 in water
Density 3.437g/mL
CAS 1302-82-5
Health Hazard 1 Danger
UN Number UN3283
Chemical Name or Material Aluminum selenide
Merck Index 14,361
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Pieces
Assay 99%
Particle Size 6.5mm & down
Sensitivity Moisture Sensitive
Quantity 1g
Health Hazard 2 GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.