missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum selenide, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 1313214

 View more versions of this product

Catalog No. AA1313214



Specifications

Specifications

Aluminum selenide
(metals basis)
UN3283
unii-4r7pmp996q, aluminum selenide, aluminium selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide
290.84
99%
Odorless
Moisture Sensitive
25g
1302-82-5
MFCD00014158
14,361
Decomposes to Al203 and SeO2 in water
Pieces
3.437g/mL
6.5mm & down
Brown

Chemical Identifiers

1302-82-5
MFCD00014158
unii-4r7pmp996q, aluminum selenide, aluminium selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

TSCA : TSCA

Documents