missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum sec-butoxide, 95%, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 4371222

 View more versions of this product

Catalog No. AA4371222



Description

Description

  • Packaged under Argon
Specifications

Specifications

Aluminum sec-butoxide
27°C (82°F)
ChemSeal™ bottles
Moisture sensitive
100g
C12H30AlO3
UN1992
2-butanol, aluminum salt, 2-butanol, aluminum salt 3:1, al o-sec-bu 3, aluminium tri-s-butoxide, aluminium tri-sec-butanolate, aluminiumtri-sec-butoxide, aluminum butan-2-olate, aluminum sec-butoxide, aluminum sec-butylate, aluminum tri-sec-butoxide
CCC(C)O.CCC(C)O.CCC(C)O.[Al]
249.351
246.32
95%
0.967 g/mL
Alcohol-like
1.439
200°C to 206°C (30mmHg)
2269-22-9
MFCD00009327
3910908
LWGPRERTOLVRLK-UHFFFAOYSA-N
aluminum;butan-2-ol
50931103
Liquid

Chemical Identifiers

2269-22-9
249.351
LWGPRERTOLVRLK-UHFFFAOYSA-N
50931103
CCC(C)O.CCC(C)O.CCC(C)O.[Al]
C12H30AlO3
MFCD00009327
2-butanol, aluminum salt, 2-butanol, aluminum salt 3:1, al o-sec-bu 3, aluminium tri-s-butoxide, aluminium tri-sec-butanolate, aluminiumtri-sec-butoxide, aluminum butan-2-olate, aluminum sec-butoxide, aluminum sec-butylate, aluminum tri-sec-butoxide
aluminum;butan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H312-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H226-H319-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 218-871-2

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

Documents