Learn More
Aluminum sec-butoxide, 95%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
$64.49 - $101.58
Chemical Identifiers
CAS | 2269-22-9 |
---|---|
Molecular Formula | C12H30AlO3 |
Molecular Weight (g/mol) | 249.351 |
MDL Number | MFCD00009327 |
InChI Key | LWGPRERTOLVRLK-UHFFFAOYSA-N |
Synonym | aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide |
PubChem CID | 50931103 |
IUPAC Name | aluminum;butan-2-ol |
SMILES | CCC(C)O.CCC(C)O.CCC(C)O.[Al] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
2269-22-9 | |
249.351 | |
LWGPRERTOLVRLK-UHFFFAOYSA-N | |
50931103 | |
CCC(C)O.CCC(C)O.CCC(C)O.[Al] |
C12H30AlO3 | |
MFCD00009327 | |
aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide | |
aluminum;butan-2-ol |
Specifications
2269-22-9 | |
200°C to 206°C (30 mmHg) | |
Alcohol-like | |
C12H30AlO3 | |
MFCD00009327 | |
UN1992 | |
Moisture sensitive | |
LWGPRERTOLVRLK-UHFFFAOYSA-N | |
aluminum;butan-2-ol | |
50931103 | |
95% | |
Aluminum sec-butoxide |
0.967 g/mL | |
27°C (82°F) | |
ChemSeal™ bottles | |
1.439 | |
100 g | |
3910908 | |
aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide | |
CCC(C)O.CCC(C)O.CCC(C)O.[Al] | |
249.351 | |
246.32 | |
Liquid |
Safety and Handling
GHS H Statement
H226-H302-H312-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H226-H319-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 218-871-2
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only