Learn More
Aluminum sec-butoxide, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03072136
Description
It is used for active catalyst and support, inorganic membrane, redrcing agent of aldehyde-ketone, oxidant of aldehyde-ketone, gel forming agent of printing ink and paint, dehydrant, water proofing agent and aluminum base grease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum sec-butoxide | |
0.967 g/mL | |
27°C (82°F) | |
C12H30AlO3 | |
MFCD00009327 | |
UN1992 | |
Moisture sensitive | |
Hydrolyzes in water; Soluble in aromatic hydrocarbons, alcohols | |
CCC(C)O.CCC(C)O.CCC(C)O.[Al] | |
249.351 | |
246.32 | |
Liquid |
2269-22-9 | |
200°C to 206°C (30 mmHg) | |
Alcohol-like | |
1.439 | |
500 g | |
3910908 | |
aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide | |
LWGPRERTOLVRLK-UHFFFAOYSA-N | |
aluminum;butan-2-ol | |
50931103 | |
95% |
Chemical Identifiers
2269-22-9 | |
249.351 | |
LWGPRERTOLVRLK-UHFFFAOYSA-N | |
50931103 | |
CCC(C)O.CCC(C)O.CCC(C)O.[Al] |
C12H30AlO3 | |
MFCD00009327 | |
aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide | |
aluminum;butan-2-ol |
Safety and Handling
GHS H Statement
H226-H302-H312-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H226-H319-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 218-871-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only