missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum sec-butoxide, 95%

Manufacturer:  Alfa Aesar™ 3072157

 View more versions of this product

Catalog No. AA3072157



Specifications

Specifications

Aluminum sec-butoxide
2269-22-9
27° (82°F)
Alcohol-like
Moisture Sensitive
C12H30AlO3
UN1992
Aluminum tri-sec-butoxide
LWGPRERTOLVRLK-UHFFFAOYSA-N
aluminum;butan-2-ol
50931103
Liquid
200°C to 206°C (30mmHg)
4 x 500g
0.967g/mL
1.4390
95%
MFCD00009327
3910908
Hydrolyzes in water; Soluble in aromatic hydrocarbons, alcohols
CCC(C)O.CCC(C)O.CCC(C)O.[Al]
249.351
246.32

Chemical Identifiers

2269-22-9
249.351
LWGPRERTOLVRLK-UHFFFAOYSA-N
50931103
CCC(C)O.CCC(C)O.CCC(C)O.[Al]
C12H30AlO3
MFCD00009327
Aluminum tri-sec-butoxide
aluminum;butan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H312-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA

SDS Documents