missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Al) Solute: Aluminum Chloride Hexahydrate Solvent: Dilute Hydrochloric Acid

Manufacturer:  Fisher Scientific Company SA442500

 View more versions of this product

Catalog No. SA442-500



 

Specifications Description & Specifications

Specifications

Certified Grade
990 to 1010ppm (by wet Method)
1
1000ppm ±1%
MFCD00134029
Lot Number Used Aluminum ref. Std.
Aluminum, Reference Standard Solution
500mL
Liquid
Colorless
1000ppm ±1%
Poly Bottle
7732-18-5,7784-13-6,7647-01-0

Chemical Identifiers

7784-13-6,7647-01-0,7732-18-5
MFCD00134029
Aluminum, Reference Standard Solution, 1000ppm ±1%
Certified Grade

Safety and Handling

SDS & Safety Documents

DOTInformation : DOT Class 8, : Corrosive