missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Powder, -325 mesh, 99% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8832222

 View more versions of this product

Catalog No. AA8832222



Specifications

Specifications

Aluminum
11145-27-0
99% (Metals basis)
MFCD00143595
CSDREXVUYHZDNP-UHFFFAOYSA-N
[Al].[Si]
55.067
Powder
100g
Silver Gray powder
AlSi
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminium silicon, aluminium-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
Odorless
aluminum;silicon
9793685
Silver to Gray

Chemical Identifiers

11145-27-0
55.067
CSDREXVUYHZDNP-UHFFFAOYSA-N
9793685
[Al].[Si]
AlSi
MFCD00143595
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminium silicon, aluminium-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
aluminum;silicon
Safety and Handling

Safety and Handling

H228-H261; Flammable solid. In contact with water releases flammable gas

SDS Documents