missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Potassium Sulfate Dodecahydrate (Powder/USP/FCC), Fisher Chemical

$482.93 - $1,110.47

Chemical Identifiers

CAS 7784-24-9
Physical Form Solid
Molecular Formula AlKO8S2 · 12 H2O
Formula Weight 474.39
MDL Number MFCD00149143
Synonym Potash Alum
Chemical Name or Material Aluminum Potassium Sulfate Dodecahydrate
Assay 99.0 to 100.5% (USP)
Grade FCC/USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
A605-212 SDS Thermo Fisher Scientific
FLA605212
Poly Bottle 2.5kg Each for $1,110.47
A605-500 SDS Fisher Scientific Company
FLA605500
Poly Bottle 500g Each for $482.93
Case of 6 Each for $2,897.58
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

7784-24-9
AlKO8S2 · 12 H2O
MFCD00149143
Aluminum Potassium Sulfate Dodecahydrate
FCC/USP
Solid
474.39
Potash Alum
99.0 to 100.5% (USP)

Specifications

92°C
2.5kg
AlKO8S2 · 12 H2O
3.0 to 3.5
0.002% max. (USP)
0.003% max. (FCC)
Pass Test (FCC)
Poly Bottle
Pass Test (USP)
0.003% max. (FCC)
Aluminum Potassium Sulfate Dodecahydrate
FCC/USP
474.39
White
Solid
3ppm max. (FCC)
43.0 to 46.0% (USP)
Potash Alum
Meets Requirements (USP)
MFCD00149143
Pass Test (FCC)
7784-24-9
99.0 to 100.5% (USP)

Safety and Handling