missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum Potassium Sulfate, 12-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor 054605
Specifications
Aluminum Potassium Sulfate | |
7784-24-9 | |
2.5 kg | |
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+] | |
474.372 | |
CHEBI:86465 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 1.73kg |
12-Hydrate | |
Crystals | |
AlH24KO20S2 | |
GNHOJBNSNUXZQA-UHFFFAOYSA-J | |
aluminum;potassium;disulfate;dodecahydrate | |
62667 | |
474.39 | |
Poly Bottle |
Chemical Identifiers
7784-24-9 | |
474.372 | |
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate | |
CHEBI:86465 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+] |
AlH24KO20S2 | |
GNHOJBNSNUXZQA-UHFFFAOYSA-J | |
62667 | |
aluminum;potassium;disulfate;dodecahydrate |