Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum plate, 6.35mm (0.25 in.) thick, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Aluminum plate
CAS 7429-90-5
Name Note 6.35mm (0.25 in.) thick
MDL Number MFCD00134029
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11062RY

  SDS
alfa aesar™
11062RY
25x25cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Plate
Avg. Mol. Wt. or Mol. Wt. Range Alloy 1100H14, Al:Cu; ≥99:0.12 wt%
TSCA TSCA
Name Note 6.35mm (0.25 in.) thick
CAS 7429-90-5
Assay 99%
Quantity 25x25cm
Chemical Name or Material Aluminum plate
Weight ≈1071g/25x25cm
MDL Number MFCD00134029
Assay Percent Notes (metals basis)