Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum, plasma standard solution, Specpure™, Al 10,000μg/mL

Chemical Identifiers

Chemical Name or Material Aluminum plasma standard solution
Name Note Al 10000μg/mL
Purity Grade Specpure™
MDL Number MFCD00134029
UN Number UN1789
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14405AD

 
alfa aesar™
14405AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14405AE

 
alfa aesar™
14405AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14405AP

 
alfa aesar™
14405AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Aluminum plasma standard solution
Quantity 50mL
TSCA TSCA
Name Note Al 10000μg/mL
Purity Grade Specpure™
Physical Form Liquid
Concentration Al in 5% HCl
MDL Number MFCD00134029
UN Number UN1789
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.