missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum, plasma standard solution, Al 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13856AE

 View more versions of this product

Catalog No. AA13856AE



Specifications

Specifications

Aluminum
Liquid
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
5359268
Liquid
100mL
7429-90-5,7429-90-5,7732-18-5
Al
Hydrochloric acid: 5.0%; Aluminum: 0.1%; Water: 94.9%
[Al]
26.982
CHEBI:33629
23 hPa (17mm Hg) at 20°C

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
CHEBI:33629
[Al]
Al
MFCD00134029
5359268
aluminum
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-595-7

Documents