missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum perchlorate nonahydrate, Reagent Grade

Manufacturer:  Alfa Aesar™ 1165922

 View more versions of this product

Catalog No. AA1165922



Specifications

Specifications

Aluminum perchlorate nonahydrate
AlCl3H18O21
UN1481
CYRDFMGMNQJULY-UHFFFAOYSA-K
aluminum;triperchlorate;nonahydrate
16211522
Crystalline
Hygroscopic
100g
81029-06-3
MFCD00149130
aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, aluminum perchlorate nonahydrate, reagent, aluminium 3+ nonahydrate triperchlorate
O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]
487.455
487.46 (325.33 Anhydrous)
Reagent
82°C

Chemical Identifiers

81029-06-3
487.455
CYRDFMGMNQJULY-UHFFFAOYSA-K
16211522
O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]
AlCl3H18O21
MFCD00149130
aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, aluminum perchlorate nonahydrate, reagent, aluminium 3+ nonahydrate triperchlorate
aluminum;triperchlorate;nonahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

TSCA : TSCA

Documents