Please login to your online account to display your discounted pricing

Aluminum oxide, White powder, NanoTek™, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Aluminum oxide
Name Note Colloidal Dispersion
CAS 1344-28-1
Physical Form Powder
Assay Percent Range 23% in H2O
Molecular Formula Al2O3
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4490822

 
alfa aesar
4490822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4490836

 
alfa aesar
4490836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA44908A3

 
alfa aesar
44908A3
2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 100g
Color White
Chemical Name or Material Aluminum oxide
Name Note Colloidal Dispersion
Assay Percent Range 23% in H2O
Molecular Formula Al2O3
MDL Number MFCD00003424
CAS 1344-28-1