Please login to your online account to display your discounted pricing

Aluminum oxide, White powder, 99% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Aluminum oxide
CAS 1344-28-1
Physical Form White powder
Assay Percent Range 99% (Metals basis)
Molecular Formula Al2O3
Formula Weight 101.96g/mol
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4465914

 
alfa aesar
44659-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Color White
Quantity 25g
Chemical Name or Material Aluminum oxide
CAS 1344-28-1
Physical Form White powder
Assay Percent Range 99% (Metals basis)
Molecular Formula Al2O3
Formula Weight 101.96g/mol
MDL Number MFCD00003424