missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide, for TLC, with gypsum

For TLC

Manufacturer:  Alfa Aesar™ 4611336

 View more versions of this product

Catalog No. AA4611336



Specifications

Specifications

Aluminum oxide
1344-28-1
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
Powder
5 to 15 Micron APS
2980°C
2045°C
With gypsum
Al2O3
14,356
[O-2].[O-2].[O-2].[Al+3].[Al+3]
101.96
101.96
Odorless
1.765
White
500g

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
Al2O3
MFCD00003424
9989226
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Safety and Handling

Safety and Handling

RTECSNumber : BD1200000

TSCA : TSCA

SDS Documents