missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide, super activated, basic, Grade I

Manufacturer:  Alfa Aesar™ 4588630

 View more versions of this product

Catalog No. AA4588630



Specifications

Specifications

Aluminum oxide
1344-28-1
Al2O3
14,356
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
Powder
Odorless
1.765
White
250g
Super activated, basic
Al2O3
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O-2].[O-2].[O-2].[Al+3].[Al+3]
101.96
101.96
Grade I
50 to 200 Micron APS
2980°C
2045°C

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents