Please login to your online account to display your discounted pricing

Aluminum Oxide Substrate, 12.7 x 12.7 x 1mm, Polished One Side, R Plane, Alfa Aesar™

$161.23 - $661.47

Chemical Identifiers

Chemical Name or Material Aluminum Oxide Substrate, R Plane
Name Note Polished One Side
Molecular Formula Al2O3
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43247KT

 
alfa aesar
43247KT
1 ea. Each for $161.23

AA43247KU

 
alfa aesar
43247KU
5 ea. Each for $661.47
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Aluminum Oxide Substrate, R Plane
MDL Number MFCD00003424
Name Note Polished One Side
Molecular Formula Al2O3