Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum Oxide Substrate, 12.7 x 12.7 x 1mm, Polished One Side, C Plane, Alfa Aesar™

$198.89 - $726.26

Chemical Identifiers

Chemical Name or Material Aluminum Oxide Substrate, C Plane
Name Note Polished One Side
Molecular Formula Al2O3
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43248KT

 
alfa aesar™
43248KT
1 ea. Each for $198.89

AA43248KU

 
alfa aesar™
43248KU
5 ea. Each for $726.26
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Aluminum Oxide Substrate, C Plane
Name Note Polished One Side
Molecular Formula Al2O3
MDL Number MFCD00003424