missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum Oxide Substrate, 10 x 10 x 1mm, Polished One Side, A Plane, Alfa Aesar™

$124.61 - $450.27

Chemical Identifiers

Molecular Formula Al2O3
MDL Number MFCD00003424
Chemical Name or Material Aluminum Oxide Substrate, A Plane, Polished One Side
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40280KT Alfa Aesar™
40280KT
1 ea. Each for $124.61
AA40280KU Alfa Aesar™
40280KU
5 ea. Each for $450.27
Specifications Description & Specifications

Chemical Identifiers

Al2O3
MFCD00003424
Aluminum Oxide Substrate, A Plane, Polished One Side

Specifications

1 ea.
Al2O3
MFCD00003424
Aluminum Oxide Substrate, A Plane, Polished One Side