Please login to your online account to display your discounted pricing

Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, R Plane, Alfa Aesar™

$123.55 - $488.19

Chemical Identifiers

Chemical Name or Material Aluminum Oxide Substrate, R Plane
Name Note Polished Two Sides
Molecular Formula Al2O3
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38500KT

 
alfa aesar
38500KT
1 ea. Each for $123.55

AA38500KU

 
alfa aesar
38500KU
5 ea. Each for $488.19
Description & Specifications

Specifications

Quantity 1 ea.
MDL Number MFCD00003424
Chemical Name or Material Aluminum Oxide Substrate, R Plane
Name Note Polished Two Sides
Molecular Formula Al2O3