Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, A Plane, Alfa Aesar™

$118.53 - $461.78

Chemical Identifiers

Chemical Name or Material Aluminum Oxide Substrate, A Plane
Name Note Polished One Side
Molecular Formula Al2O3
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38498KT

 
alfa aesar
38498KT
1 ea. Each for $118.53

AA38498KU

 
alfa aesar
38498KU
5 ea. Each for $461.78
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Aluminum Oxide Substrate, A Plane
Name Note Polished One Side
Molecular Formula Al2O3
MDL Number MFCD00003424