Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Aluminum oxide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Formula Weight 101.96g/mol
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41043KS

 
alfa aesar™
41043-KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 ea.
Quantity 1 each
Chemical Name or Material Aluminum oxide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.99%
Assay Percent Notes (metals basis)
Formula Weight 101.96g/mol
MDL Number MFCD00003424
TSCA TSCA