Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide, single crystal

Chemical Identifiers

Chemical Name or Material Aluminum oxide
CAS 11092-32-3
Name Note Single crystal, 25.4mm dia x 0.5mm thick, random orientation
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36649KG

  SDS
alfa aesar™
36649KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36649KJ

  SDS
alfa aesar™
36649KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Aluminum oxide
TSCA TSCA
Name Note Single crystal, 25.4mm dia x 0.5mm thick, random orientation
MDL Number MFCD00003424
CAS 11092-32-3
Quantity 1 piece